Hollow Cylinder
# de catalogue 470004-038
Fournisseur: Science First
Spécifications
- Description:Hollow Cylinder
- Cat. no.:470004-038
- Supplier No.:N-142
Spécifications
A propos de cet article
The Perfect accessory for an Electrostatic Generator
Use this apparatus to show students that an electric charge lies on the outer surface - not the inner surface - of a charged body.
Customers Who Bought This Also Bought