High RPM Steam Engine
# de catalogue 470005-846
Fournisseur: Jensen Manufacturing
Spécifications
- Description:High RPM Steam Engine
- Size:23 x 23 x 23 cm
- Cat. no.:470005-846
- Supplier No.:70
Spécifications
A propos de cet article
The High rpm Steam Engine features an electrically-heated horizontal boiler.
- Water-Gauge Safety Valve
- Whistle
- Throttle
- Two-Speed Pulley
- Electronically-Heated Horizontal Boiler
The apparatus includes a water-gauge safety valve, whistle, throttle, two-speed pulley, and 400W heater.
The engine may be used to drive our Electric Generator.
Ordering information: The engine includes a three-wire cord and plug for operation on 110VAC.
Customers Who Bought This Also Bought