High Power Light Source
Fournisseur: Science First
Certificats
A propos de cet article
For use with our Ripple Tank
- 150-watt
- Complete with cord, socket, bulb, and spring clamp
This light source is a clear bulb mounted inside a flat black shield with only one hole for light to escape.
The fully vented lamp shield can be rotated to provide a point source or a straight-line source. For added security and stability, we've included the Pole Cat™. This very necessary accessory will improve the grip of the Light Source.
Customers Who Bought This Also Bought