A propos de cet article
These alkaline batteries provide a much longer life than conventional carbon-zinc batteries.
- High quality
- Long storage life
- Guaranteed against leakage
- Contains no mercury or cadmium
Enhanced Rayovac High Energy alkaline batteries offer the ultimate in energy and savings. For reliability in devices you use every day, Rayovac High Energy provides power that lasts as long as the competition at a competitive price. Features Smart Seal technology, a highly engineered seal that allows more space for active ingredients.