Hero’s Engine
# de catalogue 470149-036
Fournisseur: VWR International
Spécifications
- Description:Hero’s Engine
- Cat. no.:470149-036
- Supplier No.:PH20505A
Spécifications
A propos de cet article
Students Can Simulate the Earliest Steam Engine.
- Includes a flask, chain, and rubber stopper
Suspended by a chain over a flame, this apparatus simulates the earliest form of steam engine (100 A.D.). The steam produced is ejected from the 125 ml flask’s side nozzles causing the flask to rotate.
Customers Who Bought This Also Bought