Hemastix
# de catalogue 470020-100
Fournisseur: THE LYNN PEAVEY COMPANY
Spécifications
- Description:Hemastix
- Cat. no.:470020-100
- Supplier No.:02135
- No. of Strips:50
Spécifications
A propos de cet article
Test saves time In experiments.
- Package of 50 strips
- Sample scale on bottle
Detect the peroxide-like activity of hemoglobin in a substance with these convenient test sticks. This material, commonly used at crime scenes, determines if the evidence seen is actually blood spatter, before any lab analysis is conducted. Simply apply a small amount of distilled water to the Hemastix, then rub the stick on a sample and compare the results to the chart on the bottle. If the reaction is positive, the blood sample can be collected as evidence.
Customers Who Bought This Also Bought