Helium-Neon Laser
# de catalogue 470004-978
Fournisseur: Industrial Fiber Optics
Spécifications
- Description:Helium-Neon lasers, 0.8 mW, modulated
- Cat. no.:470004-978
- Supplier No.:IF-HN08M
Spécifications
A propos de cet article
The Helium-Neon Laser is a strong laser source for holography and interferometry.
- Wavelength: 632.8 nm
- AM/FM Radio Transmission
- Solid-State Electronic Design
- Tamper-Resistant Screws for Safety
- Ideal for Holography and Interferometry
Helium neon (He Ne) lasers have wavelength stability (632.8 nm) and long coherence length. Features include AM/FM radio transmission, diffraction, and single and double slit experiments. The versatile lasers are stored in attractive blue acrylic casing.
The lasers have solid-state electronic design, a fool-proof mechanical beam stop, tamper-resistant screws for safety, threaded mounts for holding lenses; filters; and diffraction gratings, and rubber feet for adhesion on slick lab surfaces.
Customers Who Bought This Also Bought