Heavy Duty Beating Net
Catalog # 470104-932
Supplier: Cumings
Specifications
- Description:Net, Beating, Heavy Duty
- Cat. no.:470104-932
- Supplier No.:AMN
Specifications
About this item
Net is made of muslin material so even the smallest specimens will not be missed. Good for sweeping through grasses.
This beating net features a 16" Aluminum Bow and 36" Aluminum Handle.
Customers Who Bought This Also Bought