Heating Mantle Controller
# de catalogue 470202-750
Fournisseur: Glas-Col
Spécifications
- Description:Heating Mantle Controller
- Cat. no.:470202-750
- Supplier No.:104A PL312
Spécifications
A propos de cet article
Regulates Heating Mantles, Tapes and Cords, Small Furnaces, Ovens and Hot Plates.
- Analog controller
- Provides 5.5% to 100% full power
Anodized aluminum case with fuse, pilot light, and 6' grounded line cord. Size: 4¼" L x 2½" W x 2" H. For 120V, 50/60Hz, 1500W. CSA listed.
Customers Who Bought This Also Bought