Wax Marking Pencils
Catalog # 470152-050
Supplier: EATON OFFICE SUPPLY
Specifications
- Color:Red
- Description:Wax marking pencil (china marker)
- Cat. no.:470152-050
- Supplier No.:SAN 02062
Specifications
About this item
Heat Resistant.
- Great for marking porcelain, glass, or metal
Customers Who Bought This Also Bought