Ward's® Hay Medium Solution
Fournisseur: Avantor
Certificats
A propos de cet article
Similar medium to Similar to Cereal Grass medium and Cerophyll. For culture of protists.
- Sterile- no autoclaving
- Convenient - prepared for you
- Prepared consistenly every time
Concentrate comes with a vial of heat-treated wheat seeds.
Ordering information: These products contain living or perishable materials
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought