Hand-Driven Illuminated Planetarium
# de catalogue 470006-100
Fournisseur: Science First
Spécifications
- Description:Hand-Driven Illuminated Planetarium
- Cat. no.:470006-100
- Supplier No.:EL-400
Spécifications
A propos de cet article
Study different concepts utilizing sunlight or darkness.
- Demonstrates a variety of phenomena
- Sun can proviide illumination
- Includes a study guide
Use the gear-and-chain-driven planetarium in daylight to study seasons and the tides, or darken the room and the illuminated sun allows investigation of moon phases, eclipses, midnight sun, twilight, dawn, length of day, and more. A study guide with activities is included. 110 VAC with a 5' cord. Size: 20"L x 14"W.
Customers Who Bought This Also Bought