Hand Vacuum Pump
Catalog # 470006-924
Supplier: United Scientific Supplies
Specifications
- Description:Hand Vacuum Pump
- Cat. no.:470006-924
- Supplier No.:HAP001
Specifications
About this item
Dual use pump for create a vacuum or pressurizing.
- Smooth action
- Seamless stainless steel
This economical pump combines the best abilities of a pressure pump with those of a vacuum pump. When the bottom nozzle is connected to a container, it can be used as a vacuum pump. When the side nozzle is connected, it becomes a pressure pump. The “two-way” piston provides better sealing than bicycle pumps do. Cylinder is made of seamless stainless steel. Size: 20 cm L x 2.25 cm dia.
Customers Who Bought This Also Bought