Generator, DC, Hand Held
# de catalogue 470003-564
Fournisseur: United Scientific Supplies
Spécifications
- Description:Generator, DC, Hand Held
- Cat. no.:470003-564
- Supplier No.:GENH02
Spécifications
A propos de cet article
Demonstrate the transfer of mechanical energy into electrical energy with a simple motion of your hand.
- Transparent body
- Comes with two 15" leads
The Hand Held DC Generator provides the means, you provide the power. Simply turn the hand-crank to produce a terminal voltage of 6.3V and a maximum current out of 0.2A. The transparent body lets everyone see exactly what is going on inside to produce this.
Customers Who Bought This Also Bought