Halobacterium Agar
# de catalogue 470180-654
Fournisseur: Avantor
Numéro CAS:
Spécifications
- Description:Prepared Halobacterium Agar Tubes
- Cat. no.:470180-654
- Supplier No.:WARD470180-654
Spécifications
A propos de cet article
For cultivation of halophilic bacilli.
This set contains 2 tubes of Halobacterium Broth and 10 tubes of Halobacterium Agar.
Halophilic organisms like this one require a growth medium with a high salt content. As such, salt may be seen to precipitate out of the medium. This is not an indication of contamination or otherwise poor quality.
Customers Who Bought This Also Bought