Gravelometer
# de catalogue 470018-586
Fournisseur: WILDLIFE SUPPLY CO.
Spécifications
- Description:Gravelometer (Particle Size Analyzer)
- Cat. no.:470018-586
- Supplier No.:14-D40
Spécifications
A propos de cet article
For Sand, Sediment, Gravel, and More
Use this particle size analyzer to grade or measure gravel and small cobble-sized bed sediments in the field. More accurate than a ruler, the device features 14 square holes of common sieve sizes. A scale in 10mm increments is provided along one side, so the meter can measure up to 310mm. Size: 13 1/2" x 11".
Customers Who Bought This Also Bought