ContactUsLinkComponent
Gravel, Pebbles and Sand
Gravel, Pebbles and Sand
Catalog # 470145-378
PEBBLES AQUARIA - 1/2IN 5LB/PK
Gravel, Pebbles and Sand
Catalog # 470145-378
Specifications
  • Volume:
    5 lbs.
  • Color:
    Natural
  • Description:
    Pebbles
  • Cat. no.:
    470145-378
  • Supplier No.:
    10502
Specifications
About this item
Attractive Bottom Cover Enhances Natural Filtration And Nitrogen Cycling
This decorative gravel is also essential for maintaining your aquarium, acting as a substrate for the bacteria cultures necessary for a healthy tank. Requires 1 lb. of gravel per gallon.
Gravel contains no lead or chrome and won’t change the pH or chemistry of the tank. Pebble sizes up to 1/4" dia.
Customers Who Bought This Also Bought