Ward's® Live Grassfrogs (Rana pipiens)
Fournisseur: CLAUDE LAMBERT
Certificats
A propos de cet article
Grass frogs are low maintenance and seasonally available, typically from September to July.
- Great for In-Class Observation
- Low-Maintenance
- Also Known as Leopard Frogs
Keep in an aquarium or terrarium with both wet and dry areas and feed crickets and/or earthworms.
Ordering information: This kit includes living or perishable materials. They may not be available in all states. Check with your local department of agriculture.
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
CANADIAN CUSTOMERS: Please note that due to Canadian import restrictions, frogs can no longer be packed with sphagnum moss.
CANADIAN CUSTOMERS: Please note that due to Canadian import restrictions, frogs can no longer be packed with sphagnum moss.
Customers Who Bought This Also Bought