Metric Graduated Cylinders
# de catalogue 470046-296
Fournisseur: American Scientific
Small Business Enterprise
Spécifications
- Description:Metric Graduated Cylinders
- Form:Tall
- Cat. no.:470046-296
- Supplier No.:2203-10
Spécifications
A propos de cet article
These graduated measuring cylinders are made of high quality polypropylene plastic and feature an easy pour spout. Extra-wide hexagon base helps to prevent spilling. Set includes 10 ml, 25 ml, 50 ml, 100 ml, 250 ml, 500 ml and 1,000 ml cylinders. Set of seven.
Customers Who Bought This Also Bought