Somso® Gorilla Skull
Fournisseur: MARCUS SOMMER
Certificats
A propos de cet article
Compare the structural differences between male and female gorillas (Gorilla gorilla), noting the smaller overall size, and size of central crest and brow ridges, of the female, by using this unmounted plastic, adult female skull with the SOMSO male gorilla skull. Both feature a removable lower jaw.
- Cast from real specimens
- Male and female versions available for comparison
- 5 year manufacturers warranty
Customers Who Bought This Also Bought