Glossopteris sp. (Permian)
# de catalogue 470027-546
Fournisseur: ADAM MOORE MO
Spécifications
- Description:Glossopteris sp
- Cat. no.:470027-546
- Supplier No.:535795
Spécifications
A propos de cet article
Leaf impression on matrix, widely used in correlation studies. Average size: 3" x 3". Australia.
Customers Who Bought This Also Bought