Ward's® Live Gloeocapsa Culture
Fournisseur: Ward's Science
Certificats
A propos de cet article
This colonial cyanobacteria is structured as several cells bound together in a gelatinous "mucilaginous sheath".
Commonly found in masses on wet rocks in aquatic or semi-aquatic habitats.
Genus: Gloeocapsa Species: Not specified
Customers Who Bought This Also Bought