Glass Tubing Manipulator
Catalog # 470016-798
Supplier: JET RUBBER COMPANY
Specifications
- Description:Glass Tubing Manipulator, Pair
- Cat. no.:470016-798
- Supplier No.:6199000
Specifications
About this item
Protects hands by eliminating accidents when inserting glass tubing into cork or rubber stoppers.
- Made from hard rubber with large shield in front
- Overall diameter is 76 mm
Customers Who Bought This Also Bought