Glass Tubing Cutter
# de catalogue 470019-484
Fournisseur: C & A Scientific
Spécifications
- Description:Glass Tubing Cutter
- Material:Stainless Steel
- Cat. no.:470019-484
- Supplier No.:97-1601
Spécifications
A propos de cet article
Made for easy glass cutting.
- Cuts up to 11/2" dia. glass tubes
The spring loaded cutting wheel stays open until used.
Customers Who Bought This Also Bought