Student Grade Glass Slide and Coverslip Lab Pack
# de catalogue 470018-998
Fournisseur: Avantor
Spécifications
- Description:Glass slides and coverslip
- Cat. no.:470018-998
- Supplier No.:WARD470018-998
Spécifications
A propos de cet article
These are student grade microscope slides.
The set includes everything necessary to prepare a dozen microscope slides: 1" x 3" glass slides, 22 mm square coverslips, slide labels, and a clear plastic slide holder.
Customers Who Bought This Also Bought