Glass Burets with Plastic Stopcock
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Made of high-quality, Leakproof glass, these burets are perfect for student use.
- White-Enameled Graduations
- Burets Are Individually Calibrated
- Leakproof Glass Construction
- Removable Stopcocks Included
- Burets Resist Most Chemicals
The burets resist most chemicals and are individually calibrated.
The plastic stopcocks require no lubrication, provide excellent flow control, and can be removed for easy cleaning.
Customers Who Bought This Also Bought