Ward's® Giant Toad (Bufo marinus)
Fournisseur: BRUCE EDELMAN
Certificats
A propos de cet article
This toad thrives with minimum attention and may be housed in a terrarium with access to water and dry land.
- Low maintenance
- Excellent for physiology studies
- Exhibits territoriality and defensive behavior
Feed live insects such as crickets.
Size ranges from 10 to 13 cm.
Ordering information: These products contain living or perishable materials
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought