Geology Specimen Storage Trays
# de catalogue 470022-346
Fournisseur: Lane Science
Spécifications
- Description:Geology Storage Trays
- Cat. no.:470022-346
- Supplier No.:301-T
Spécifications
A propos de cet article
Designed for Geology Specimen Cabinet
Use these drawers that feature easy-glide edges, two heavy-gauge bar handles, and a large label holder with our Geology Specimen Cabinet. The reinforced steel will not sag under the weight of geological specimens. Size: 25"W x 30"D x 1 7/8"H; weight: 11 lbs.
Customers Who Bought This Also Bought