Geology Specimen Cabinet
# de catalogue 470022-344
Fournisseur: Lane Science
Spécifications
- Description:Geology specimen cabinet
- Cat. no.:470022-344
- Supplier No.:301|LANE SCIENCE EQUIPMENT CORP
Spécifications
A propos de cet article
Rugged, welded-steel construction and specialized features make this specimen cabinet the most durable choice for storing valuable scientific collections.
- Weight: 134 lbs.
- Size: 29W×32D×36⁷/₈"H
- It will accept up to 16 storage trays
- Powder-paint finish
The cabinet features a unique 'lift off' door that’s easily removed from its hinges to enable transfer of many drawers at a time. The door also features a elastomeric seal for airtight storage, locking handle, and chrome label holder.
Ordering information: Trays must be ordered separately (470022-346).
Customers Who Bought This Also Bought