Generalized Plant Cell Slide
Catalog # 470177-554
Supplier: Avantor
Specifications
- Description:Generalized Plant Cell Slide
- Cat. no.:470177-554
- Stain:Quadruple stained
- Supplier No.:WARD470177-554
- Orientation:Section
Specifications
About this item
Investigate plant cell traits
- Identifying features clearly distinguishable
- Rigorous quality control standards
Cell wall, nucleus, nucleolus, large vacuole, and chloroplasts are shown. Study guide included.
Customers Who Bought This Also Bought