General Science Student Lab Notebook
# de catalogue 470023-066
Fournisseur: MACMILLAN HOLDINGS, LLC
Spécifications
- Description:General Science Student Lab Notebook
- Cat. no.:470023-066
- No. of Pages:70
- Length:22 cm
- Width:28 cm
- Supplier No.:193088284X
Spécifications
A propos de cet article
Doubles as Lab Workbook and Reference
Geared toward general science courses, this notebook is permanently side bound, with a wrap-around back cover, that is used between the carbonless sets to prevent bleed-through. The duplicate carbonless sets allow instructor and student to each have a copy of completed work. And both sheets are micro perforated for easy tear out. For convenience, it also contains a full color periodic table, quick reference information, and safety guidelines.
Customers Who Bought This Also Bought