Genecon Activity Set with Manual
# de catalogue 470111-188
Fournisseur: NADA SCIENTIFIC LTD.
Small Business Enterprise
Spécifications
- Description:Genecon activity set with manual
- Cat. no.:470111-188
- Supplier No.:B-2637
Spécifications
A propos de cet article
Covers all the major topics in electricity. Activity manual includes 24 hands-on investigations for grades 4 and beyond.
Your students will get a 'charge' out of this kit! Includes two Genecons and 44 accessories which allow students to experience such phenomena as energy transformations, series and parallel circuits, electromagnetism, the motor effect, electrolysis of water, electroplating, wet cells, short circuits and fuses and storage cells.
Customers Who Bought This Also Bought