Gastop for Teck™ Gas Jets
Supplier: TSUNAMI TECHNOLOGY GROUP INC.
Certificates
About this item
Improve safety and prevent vandalism with these innovative caps, which lock on to most gas jets and can only be removed with the supplied key.
- Gastop slips over hose barb, providing a vandal-proof seal
- Seal against leakage
- Prevent items from being inserted into gas jet
- Unlocks with key
Designed to prevent accidental leaks, the caps are certified by the Canadian Gas Association and the American Gas Association. Please note: these gastops are made for use with Delta Faucet's Teck (tm) gas jets.
Customers Who Bought This Also Bought