Gastop for Teck™ Gas Jets
# de catalogue 470146-712
Fournisseur: Avantor
Spécifications
- Description:Gastop
- Supplier No.:GASTOP
Spécifications
A propos de cet article
Improve safety and prevent vandalism with these innovative caps, which lock on to most gas jets and can only be removed with the supplied key.
Designed to prevent accidental leaks, the caps are certified by the Canadian Gas Association and the American Gas Association. Please note: these gastops are made for use with Delta Faucet's Teck (tm) gas jets.
Customers Who Bought This Also Bought