Gas Collecting Tube
# de catalogue 470102-268
Fournisseur: ROCHESTER APPAREL & AD SPECIALTIES
Spécifications
- Description:Gas Collecting Tube
- Cat. no.:470102-268
- Supplier No.:47519-00
Spécifications
A propos de cet article
Unique, Student-Friendly Gas Collecting Device
- Safe and easy
This Pyrex® tube makes collecting and analyzing any gaseous product over water safe and easy. Developed by Mary Gallup. The tube measures 2 mm inner diameter, 2 mm wall, 177.8 mm long. Package of 10.
Customers Who Bought This Also Bought