Galapagos Tortoise Skull
# de catalogue 470029-288
Fournisseur: SKULLS UNLIMITED
Spécifications
- Description:Galapagos Tortoise Skull
- Height:3"
- Cat. no.:470029-288
- Length:6"
- Width:4"
- Supplier No.:TQ-56
- No. of Parts:1