Funnel Support
# de catalogue 470201-934
Fournisseur: Bennett Wood Specialties
Spécifications
- Description:Funnel Support, 41 cm
- Cat. no.:470201-934
- Supplier No.:2605
Spécifications
A propos de cet article
Designed to support four funnels, this well made, hardwood support attaches to any 13 mm (1/2" ) diameter rod. The wood is slotted to facilitate insertion and removal of funnels. Complete with metal clamps and aluminum thumb screw. Overall length is 41 cm.
Customers Who Bought This Also Bought