Funnel Support
# de catalogue 470123-524
Fournisseur: VWR International
Spécifications
- Description:Funnel Support, Length 16"
- Material:Hardwood
- Cat. no.:470123-524
- Supplier No.:CH0689D
Spécifications
A propos de cet article
With iron clamp for support rods of any size to 5/8". The four holes are beveled and slotted and are spaced at 41/2" center distance.
Customers Who Bought This Also Bought