Fundamantals of Physics Ballistic Pendulum
Catalog # 470018-434
Supplier: VWR International
Specifications
- Description:Ballistic Pendulum
- Cat. no.:470018-434
- Supplier No.:BLP-101
Specifications
About this item
The Ballistic Pendulum is spring-loaded for easy investigations.
- Three Firing Positions
- Pendulum Catch
- Spring-Loaded Launcher
- Horizontal Launcher
- Students Research Conservation of Momentum and Projectiles
Students can research the conservation of momentum and projectiles with this spring-loaded launcher, which features three firing positions and a pendulum catch. Users can quickly attach a photogate to measure the actual velocity of the marble. It also works as horizontal launcher.
A photogate is available separately.
Customers Who Bought This Also Bought