Fundamental Structure Lattice Model Set
# de catalogue 470029-194
Fournisseur: Klinger
Spécifications
- Description:Fundamental Structure Lattice Model Set
- Cat. no.:470029-194
- Supplier No.:7996
Spécifications
A propos de cet article
Full Form Crystal Structures Instead of Just One Cell
These nine models, constructed of wooden beads and metal rods, include examples from three major crystal symmetry systems representing many key mineral forms in greater detail by showing more of their structure. Different atoms are represented by various bright colors. Each model is clearly labeled, and the set comes with an illustrated instruction booklet. Contents: Rhombohedral structure: calcite; hexagonal structure: graphite, magnesium, and wurtzite; cubic structure: carbon dioxide, cesium chloride, copper, diamond, and sodium chloride. Size: 7–15".
Customers Who Bought This Also Bought