Spécifications
- Description:FUN-damental Chemical Reactions Kit
- No. of Lab Groups:10
- Cat. no.:470152-972
- Includes:Instructions
- Supplier No.:WARD470152-972
Spécifications
A propos de cet article
Teach Students to Recognize the Common Distinguishing Phenomena That Occur During Chemical Reactions
- Ideal for grades 7-10
- Perform synthesis, decomposition, single/double displacement and neutralization reactions
Students will learn to recognize each reaction by observing qualities such as effervescence, color change, temperature change and the formation of precipitate. By performing these experiments, students will gain an understanding of the basic characteristics of these fundamental chemical processes. Designed by science teacher Joe Gilbert.
Materials Included:
crucibles
microfuge tubes
pipets
test tubes
tubing
stoppers
bromothymol blue
phenolphthalein
wide range pH indicator
baking soda
magnesium metal
copper II sulfate
copper II chloride
sodium carbonate
ammonium hydroxide
hydrochloric acid
vinegar
microtube holder