Fuming Hot Plate
# de catalogue 470020-328
Fournisseur: SIRCHIE
Spécifications
- Description:Fuming Hot Plate
- Cat. no.:470020-328
- Supplier No.:L-9007
Spécifications
A propos de cet article
Accelerate the Development of Cyanoacrylate
Simply place this hot plate inside your fuming chamber and add 5-10 drops of cyanoacrylate into the aluminum tray. Prints will then develop in a matter of minutes, saving hours of lab time. 100 VAC.
Customers Who Bought This Also Bought