Fraction Stax Set
# de catalogue 470046-944
Fournisseur: CARSON-DELLOSA PUBLISHING CO.
Spécifications
- Description:Fraction Stax, Set/51
- Cat. no.:470046-944
- Supplier No.:ID7749
Spécifications
A propos de cet article
A colorful set of 51 plastic stacking pieces representing one whole, halves, thirds, fourths, fifths, sixths, eighths, tenths, and twelfths.
- Understanding fractions
- Three-dimensional model
The Fraction Stax is a three-dimensional model that demonstrates and teaches fraction equivalences in a tangible, easy-to-understand method.
The Fraction Stax is great for teacher demonstrations, group work, and individual studies.
Includes one nine-peg base and a Teacher's Guide.
Dimensions: 14 x 2 3/4 (35.5cm x 7cm) base. The "whole" Stax is 4 1/2 (11.5cm).
Customers Who Bought This Also Bought