ContactUsLinkComponent
Ward's® Four-Step Tumbling Abrasives
Ward's® Four-Step Tumbling Abrasives
# de catalogue 470157-402
Fournisseur:  LORTONE, INC.
FOUR STEP TUMBLING ABRASIVE KIT
Ward's® Four-Step Tumbling Abrasives
# de catalogue 470157-402
Fournisseur:  LORTONE, INC.
Spécifications
  • Description:
    Four-Step Tumbling Abrasives
  • Supplier No.:
    590-031
Spécifications
A propos de cet article
All the materials you need to grind and finish up to 14 lbs. of material are contained in this one kit.
  • Four-step kit
  • Contains silicon carbide, prepolish, and polish
Kit contains 14 oz. coarse silicon carbide, 14 oz. fine silicon carbide, 12 oz. prepolish, and 12 oz. polish.
Customers Who Bought This Also Bought