ContactUsLinkComponent
Four Stroke Petrol Engine
Four Stroke Petrol Engine
# de catalogue 470014-350
Fournisseur:  VWR International
FOUR STROKE PETROL ENGINE CONNECTS TO 3V
Four Stroke Petrol Engine
# de catalogue 470014-350
Fournisseur:  VWR International
Spécifications
  • Description:
    Four Stroke Petrol Engine
  • Size:
    8¼ x 8¼ x 14½"
  • Cat. no.:
    470014-350
  • Supplier No.:
    PH0488
Spécifications
A propos de cet article
Sized For Student Use.
  • Modeled after a typical air cooled engine
The workings of an engine are clearly explained through observing this representation four stroke petrol engine. The operation of the valves is clearly evident with the cams being driven by a gear train from the rear of the main crank. A simulated spark plug uses a small lamp to indicate the firing point and a cross section of the carburetor is shown. Mounted on a base, the engine provides screw terminals for connection to a 3V battery.
Customers Who Bought This Also Bought