Ward's® Fossil Display Pack
# de catalogue 470026-756
Fournisseur: Avantor
Spécifications
- Description:Fossil Display Pack
- Cat. no.:470026-756
- Supplier No.:WARD470026-756
- No. of Parts:8 samples
Spécifications
A propos de cet article
Eight High-Quality Fossils for Introductory Classes
- Introduces students to different Phyla of ancient life
- Keycard provides information on each fossil
A fine display set of basic fossil types for beginning geology students, this set is mounted on a study display card with information outlining each fossil and providing key information on the groups represented.
Size: 4 5/8"L x 3 5/8"W x 1 3/8"H.
Contents:8 unnumbered fossilsFossils:brachiopod, bryozoan, crinoid stem, gastropod, horn coral, petrified wood, shark tooth, trilobite.
Customers Who Bought This Also Bought