Forward Vision™ Four Student Workstations
Supplier: Diversified Woodcrafts
Certificates
About this item
All in one: classroom table and lab bench. The ForwardVision™ workstation allows up to four students to work at a single station while all are facing the teacher.
- 1" solid epoxy resin surface
- Constructed of solid oak and oak veneers
- Rubber base molding and durable chemical resistant UV finish
- All doors and drawers with lock
- Two models available: door cabinet with two adjustable shelves or the door/drawer cabinet with one adjustable shelf and a drawer
This popular workstation meets the needs of students and teachers today. The unique design leaves enough room to store four stools (not included) under the workstation.
Sink dimensions: 22³/₈W×15L×17" D
Certifications: SEFA compliant and MAS Certified Green.
Delivery: Unit comes with a sink, two combination water/gas fixtures, two GFI electrical outlets and AC duplex receptacles, and ample storage behind two locking door/drawer cabinets with one adjustable shelf and a drawer. The 2916KLH and 2936KLH include two rod sockets and LabHands for holding laptop computers and books.
Specifications
- Catalog No:
- 470107-562
- 470107-558
- 470107-564
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green