Forensic Enhancement Techniques
# de catalogue 470128-672
Fournisseur: Edvotek
Spécifications
- Description:Forensic Enhancement Techniques
- No. of Lab Groups:10
- Cat. no.:470128-672
- Supplier No.:194
Spécifications
A propos de cet article
Reveal hidden biological evidence.
- Case-based scenario
- Requires an estimated 35 minutes class/lab time
- Store refrigerated
Trace amounts of blood are often sufficient to identify the individual responsible for any number of crimes, including murder, burglary, or assault. Enhancement procedures can make a small stain of body fluid or tissue visible to the naked eye. In this experiment, students will act as detectives following the aftermath of a drug bust involving gang warfare over territory. Reagents that are routinely used as a first screen will be utilized to detect simulated blood and DNA. In addition, biological materials will be recovered from splatters, blood trajectory, and small droplets of simulated human materials.
All you need is a UV light, gloves, paper towel, and face masks. A fume hood is optional.
Ordering information: Kit includes instructions, simulated blood, leucocrystal violet solution, luminol solution, spray bottle, transfer pipets, microtest tubes.
Customers Who Bought This Also Bought