Forensic Chemistry : Fabric and Fiber Analysis Activity
# de catalogue 470200-662
Fournisseur: Kemtec
Spécifications
- Description:Forensic Chemistry : Fabric and Fiber Analysis Activity
- No. of Lab Groups:6
- Cat. no.:470200-662
- Supplier No.:10-534
Spécifications
A propos de cet article
Test a Variety of Samples
- Grade Level 10-12
- Case-based Scenario
In the Forensic Chemistry: Fabric and Fiber Analysis Activity, students use a microscope you provide and a variety of chemicals to test fibers and use them as evidence in solving a crime.
Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Customers Who Bought This Also Bought