Foraminifera Thin Section
# de catalogue 470026-822
Fournisseur: Avantor
Spécifications
- Description:Foraminifera thin section
- Supplier No.:WARD470026-822
Spécifications
A propos de cet article
Cross section of a Pennsylvanian fusulinid in chert that shows random orientations of structural features, including the proloculus and chamber walls.
Customers Who Bought This Also Bought